missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, contains no preservative, >99.8%, Solstice

Catalog No. 6001681
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001681 4 x 4 L Poly-coated Bottle
6001680 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001681 Supplier Solstice Supplier No. AH3404PC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Absorbance 0.18 AU max. at 250 nm, 0.02 AU max. at 300 nm
CAS Max % 100.0000%
Density 0.8892 g/cm3
Quantity 4 x 4 L
Flash Point −14°C
Packaging Poly-coated Bottle
UN Number UN2056
Water 0.03% max.
Beilstein 102391
Boiling Point 66°C
Solubility Information Miscible in Water at 20°C
Formula Weight 72.11
Percent Purity >99.8%
Grade HPLC
Chemical Name or Material Tetrahydrofuran, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, contains no preservative, >99.8%
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Tetrahydrofuran
EINECSNumber 203-726-8