missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran ACS AR, Macron Fine Chemicals™
macron
$6980.11 - $6980.11
Chemical Identifiers
| CAS | 109-99-9 |
|---|---|
| Molecular Formula | C4H8O |
| Molecular Weight (g/mol) | 72.11 |
| MDL Number | MFCD00005356 |
| InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
| Synonym | tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano |
| PubChem CID | 8028 |
| ChEBI | CHEBI:26911 |
| IUPAC Name | oxolane |
| SMILES | C1CCOC1 |
Chemical Identifiers
| 109-99-9 | |
| 72.11 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| 8028 | |
| oxolane |
| C4H8O | |
| MFCD00005356 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| CHEBI:26911 | |
| C1CCOC1 |
Specifications
| 109-99-9 | |
| Steel Pail | |
| MFCD00005356 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| oxolane | |
| 8028 | |
| 72.11 | |
| Tetrahydrofuran |
| 20 L | |
| C4H8O | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| C1CCOC1 | |
| 72.11 | |
| CHEBI:26911 | |
| ACS Reagent |