missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran ACS AR, Macron Fine Chemicals™

Catalog No. MK849820
Change view
Click to view available options
Quantity:
20 L
Packaging:
Steel Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK849820 20 L Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK849820 Supplier Avantor Supplier No. 849819

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Quantity 20 L
Packaging Steel Pail
Formula Weight 72.11
Grade ACS
Chemical Name or Material Tetrahydrofuran