missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, 99%, stab. with 250-350ppm BHT

Catalog No. AAL13304AU
Change view
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13304AU 1000 mL
AAL13304AP 500 mL
AAL133040F 2500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL13304AU Supplier Thermo Scientific Chemicals Supplier No. L13304AU

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 109-99-9 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrahydrofuran is used as a solvent in Grignard reagent formation processes, pharmaceutical steroids and in the preparation of organometallic reagents. It is used for HPLC, spectrophotometry and environmental testing. It is used as a resin solvent in flexographic inks for plastics coating deposition for audio and video tapes. It is used in PVC and CPVC pipe cements, polyurethane coatings and in PVC film castings.

Solubility
Miscible with water.

Notes
Highly flammable. Oxidizes readily in air to form unstable peroxides. THF exhibits high volatility and an extremely low freezing point. Keep the container tightly closed and sealed until ready for use.

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C
Density 0.889
Quantity 1000 mL
Flash Point −17°C (1°F)
Odor Ether-like
Refractive Index 1.407
UN Number UN2056
Beilstein 102391
Merck Index 14,9211
Boiling Point 66°C
Solubility Information Miscible with water.
Formula Weight 72.11
Percent Purity 99%
Chemical Name or Material Tetrahydrofuran, stabilized with 250 to 350ppm BHT
Show More Show Less
Hazard Category H225-H302-H319-H335-H336-H351
Hazard Statement GHS H Statement
H225-H351-H319-H335
Highly flammable liquid and vapor.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280i-P281-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: TETRAHYDROFURAN
EINECSNumber 203-726-8
RTECSNumber LU5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only