missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, 99.9% min., MilliporeSigma™
Specifications
Specifications
| Color | Colorless |
| Quantity | 100 mL |
| Physical Form | Liquid |
| Assay Percent Range | 99% min. |
| Linear Formula | -CH2(CH2)3O- |
| Formula Weight | 72.11 |
| Grade | NF/USP |
| Chemical Name or Material | Tetrahydrofuran, OmniSolv™ No Stabilizer |
Safety and Handling
| Recommended Storage | Room Temperature |