missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, 99.9% min., MilliporeSigma™

Catalog No. MTX02797
Change view
Click to view available options
Quantity:
100 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MTX02797 100 mL
MTX02796 1 L
MTX02791 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MTX02797 Supplier Technidata Supplier No. TX02797

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Color Colorless
Quantity 100 mL
Physical Form Liquid
Assay Percent Range 99% min.
Linear Formula -CH2(CH2)3O-
Formula Weight 72.11
Grade NF/USP
Chemical Name or Material Tetrahydrofuran, OmniSolv™ No Stabilizer
Recommended Storage Room Temperature