Learn More
Tetrahydrofuran, 99.9%, extra pure, anhydrous, stabilized with BHT
Tetrahydrofuran, >-99.85%, C4H8O, CAS Number-109-99-9, 128-37-0, tetraidrofurano, furanidine, tetrahydrofuranne, butylene oxide, tetrahydrofuran, furan, tetrahydro, oxacyclopentane, hydrofuran, 1,4-epoxybutane, tetramethylene oxide, 1L, 99.9%, 17, 10, CHEBI:26911, Undesignated, 0.8800g/mL | CAS: 109-99-9 | C4H8O | 72.11 g/mol
$257.43 - $3106.25
Chemical Identifiers
| CAS | 109-99-9 |
|---|---|
| Molecular Formula | C4H8O |
| Molecular Weight (g/mol) | 72.11 |
| MDL Number | MFCD00005356 |
| InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
| Synonym | tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano |
| PubChem CID | 8028 |
| ChEBI | CHEBI:26911 |
| IUPAC Name | oxolane |
| SMILES | C1CCOC1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC181500010
|
Thermo Scientific Chemicals
181500010 |
1 L | Glass Bottle |
Each for $257.43
|
|
||||
|
AC181500025
|
Thermo Scientific Chemicals
181500025 |
2.5 L | Glass Bottle |
Each for $562.96
|
|
||||
|
AC181500250
|
Thermo Scientific Chemicals
181500250 |
25 L | Metal Drum |
Each for $3,106.25
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 109-99-9 | |
| 72.11 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| 8028 | |
| oxolane |
| C4H8O | |
| MFCD00005356 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| CHEBI:26911 | |
| C1CCOC1 |
Specifications
| 109-99-9 , 128-37-0 | |
| 1 L | |
| Glass Bottle | |
| 1.4060 to 1.4080 (20°C, 589nm) | |
| C4H8O | |
| 0.006% max. (Coulometric) | |
| 01,1140; 02,398; 03,278; 05,649; 06,570 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| C1CCOC1 | |
| 72.11 | |
| CHEBI:26911 | |
| ≥99.85% | |
| Tetrahydrofuran, Anhydrous, Stabilized |
| 0.8800g/mL | |
| Authentic | |
| Liquid | |
| 99.9% | |
| MFCD00005356 | |
| 17, 10 | |
| 15, 9356 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| oxolane | |
| 8028 | |
| 72.11 | |
| Extra Pure |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only