missing translation for 'onlineSavingsMsg'
Learn More

Tétrahydrofurane, 99,9%, extra sec, anhydre, SC, AcroSeal™, Thermo Scientific Chemicals

Numéro de catalogue. AC610921000
Modifier l'affichage
Click to view available options
Quantité:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
AC610921000 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. AC610921000 Fournisseur Thermo Scientific Chemicals Code fournisseur 610921000

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

 

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-99-9 , 128-37-0
Formule moléculaire C4H8O
Poids moléculaire (g/mol) 72.11
Numéro MDL MFCD00005356
Clé InChI WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonyme tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
Nom de l’IUPAC Oxolane
SOURIRES C1CCOC1

Specifications

Nom ou substance chimique Tetrahydrofuran
Nom Remarque Eau < 50 ppm
Densité 0.8800g/mL
Quantité 100 mL
Désignation Test Réussi
Fourchette de pourcentages d’essai 99.9%
Pression de vapeur Pression de vapeur : 200 mbar à 20 °C
Eau 50 ppm max.
Beilstein 17, 10
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Indice de Merck 15, 9356
Poids de formule 72.11
Pourcentage de pureté ≥99.9%
Niveau Extra sec
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Precautionary Statement GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Keep container tightly closed.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Usage réservé à la recherche