missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Puriss. p.a., contains 250 ppm BHT as inhibitor, ACS Reagent, Reag. Ph. Eur., ≥99.9%, Honeywell Riedel-de Haën™

Catalog No. p-7120867
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
Packaging:
Glass Bottle
N/A
Catalog No. Quantity Packaging
60047702 1 L Glass Bottle
60047703 2.5 L Glass Bottle
60047705 4 x 2.5 L N/A
3 options

Catalog No. 60047702

Supplier: Honeywell Chemicals 873681L

Only null left

Puriss. p.a., contains 250 ppm BHT as inhibitor, ACS Reagent, Reag. Ph. Eur.,≥99.9%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C
Color Colorless
Density 0.887g/mL at 20°C, 0.889g/mL at 25°C
Boiling Point 65°C (1013 hPa)
Quantity 1 L
Explosion Limit 0.118
Flash Point −17°C (1.4°F)
Ignition Point 610°F
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 114mmHg (15°C), 143mmHg (20°C)
UN Number UN2056
Beilstein 102391
Formula Weight 72.11g/mol
Percent Purity ≥99.9%
Chemical Name or Material Tetrahydrofuran
Show More Show Less
Hazard Statement P210-P280-P301 + P312 + P330-P305 + P351 + P338-P370 + P378-P403 + P235
DOTInformation 3/PGII
RTECSNumber 203-726-8