missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™
CAS: 109-99-9 | C4H8O | 72.11 g/mol
Supplier: Thermo Scientific Chemicals 457071000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tetrahydrofuran | |
| 109-99-9,128-37-0 | |
| 66.0°C | |
| −21°C | |
| 1.407 | |
| C4H8O | |
| 0.55 mPa.s (20°C) | |
| Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone | |
| C1CCOC1 | |
| 72.107 | |
| CHEBI:26911 | |
| 99.85% |
| -108.4°C | |
| 0.8800g/mL | |
| 100 mL | |
| AcroSeal™ Glass Bottle | |
| 0.88 | |
| MFCD00005356 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| oxolane | |
| 8028 | |
| 72.11 | |
| Extra Dry |
Chemical Identifiers
| 109-99-9 | |
| MFCD00005356 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| CHEBI:26911 | |
| C1CCOC1 |
| C4H8O | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| 8028 | |
| oxolane |
Safety and Handling
GHS H statement
May cause respiratory irritation
Causes serious eye irritation
Highly flammable liquid and vapor
May form explosive peroxides
Suspected of causing cancer
Suspected of causing cancer
GHS P statement
Keep away from heat/sparks/open flames/hot surfaces
No smoking
No smoking
Wear protective gloves/protective clothing/eye protection/face protection
Avoid breathing dust/fume/gas/mist/vapors/spray
If in eyes; rinse cautiously with water for several minutes
Remove contact lenses, if present and easy to do. Continue rinsing. Wash with plenty of soap and water
Avoid breathing dust/fume/gas/mist/vapors/spray
If in eyes; rinse cautiously with water for several minutes
Remove contact lenses, if present and easy to do. Continue rinsing. Wash with plenty of soap and water
Keep container tightly closed
Danger
EINECSNumber : 203-726-8
RUO – Research Use Only