missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, 99.85%, Extra Dry, Unstabilized, AcroSeal™

CAS: 109-99-9 | C4H8O | 72.11 g/mol

Supplier:  Thermo Scientific Chemicals 457070025

 View more versions of this product

Catalog No. AC457070025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetrahydrofuran
109-99-9,128-37-0
66.0°C
−21°C
0.88
MFCD00005356
0.005% max.
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
C1CCOC1
72.11
CHEBI:26911
99.85%
-108.4°C
0.8800g/mL
2.5 L
1.407
C4H8O
0.55 mPa.s (20°C)
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
Extra Dry

Chemical Identifiers

109-99-9
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
CHEBI:26911
C1CCOC1
C4H8O
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement Highly flammable liquid and vapor. Harmful if swallowed. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May form explosive peroxides.

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

Danger

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only