missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, 99.8%, for biochemistry, unstabilized, AcroSeal™

CAS: 109-99-9 | C4H8O | 72.11 g/mol

$193.43 - $193.43

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC450530010
SDS View Documents
Thermo Scientific Chemicals
450530010
1 L AcroSeal™ Glass Bottle
Each for $193.43
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-99-9
72.11
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
CHEBI:26911
C1CCOC1

Specifications

-108.4°C
0.8800g/mL
1 L
AcroSeal™ Glass Bottle
99.8%
C4H8O
0.55 mPa.s (20°C)
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
C1CCOC1
72.11
CHEBI:26911
99.8%
Tetrahydrofuran
109-99-9,128-37-0
66.0°C
−21°C
1.407
0.88
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
Biochemical
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only