missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, 99.5% min., ACS, MilliporeSigma™
Supplier: Technidata TX028020
Specifications
| Tetrahydrofuran | |
| 109-99-9 | |
| Liquid | |
| C4H8O | |
| MFCD00005356 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| oxolane | |
| 8028 | |
| 72.11 |
| For ACS analysis | |
| 200 L | |
| 99.5%, min. | |
| -CH2(CH2)3O- | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| C1CCOC1 | |
| 72.11 | |
| CHEBI:26911 | |
| ACS |
Chemical Identifiers
| 109-99-9 | |
| 72.11 | |
| WYURNTSHIVDZCO-UHFFFAOYSA-N | |
| 8028 | |
| oxolane |
| C4H8O | |
| MFCD00005356 | |
| tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
| CHEBI:26911 | |
| C1CCOC1 |
Safety and Handling
Recommended Storage : Room Temperature