missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-3-sulfonyl chloride, Thermo Scientific™

Catalog No. p-4391478
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC76003DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC76003DA Supplier Thermo Scientific Chemicals Supplier No. CC76003DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1207346-29-9
Molecular Formula C4H7ClO3S
Molecular Weight (g/mol) 170.607
InChI Key MVZIUOJNVVIOEY-UHFFFAOYSA-N
Synonym tetrahydrofuran-3-sulfonyl chloride, chlorooxolan-3-ylsulfone
PubChem CID 49762850
IUPAC Name oxolane-3-sulfonyl chloride
SMILES C1COCC1S(=O)(=O)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 170.61
Percent Purity 97%
Chemical Name or Material Tetrahydrofuran-3-sulfonyl chloride
Hazard Category GHS Signal Word: Danger