missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran-2,3,4,5-tetracarboxylic Acid 98.0+%, TCI America™

Catalog No. p-7136528
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T097525G 25 g
T0975500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T097525G Supplier TCI America Supplier No. T097525G

Chemical Identifiers

CAS 26106-63-8
Molecular Formula C8H8O9
Molecular Weight (g/mol) 248.14
MDL Number MFCD00005364
InChI Key UFOIOXZLTXNHQH-UHFFFAOYNA-N
Synonym tetrahydrofuran-2,3,4,5-tetracarboxylic acid, tetrahydrofuran-2,3,4,5-tetracarboxylicacid, acmc-209goc, 2,5-anhydro-3,4-dicarboxy-3,4-dideoxyhexaric acid, tetrahydrofuran-2,3,4,5-tetracarboxylic acid, mixed isomers, tetrahydro-2,3,4,5-furantetracarboxylic acid, 2,3,4,5-furantetracarboxylicacid, tetrahydro
PubChem CID 97421
IUPAC Name oxolane-2,3,4,5-tetracarboxylic acid
SMILES OC(=O)C1OC(C(C1C(O)=O)C(O)=O)C(O)=O

Specifications

Melting Point 205°C
Color White
Quantity 25 g
Formula Weight 248.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrahydrofuran-2,3,4,5-tetracarboxylic Acid