missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran-2,3,4,5-tetracarboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 205°C |
| Color | White |
| Quantity | 25 g |
| Formula Weight | 248.14 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tetrahydrofuran-2,3,4,5-tetracarboxylic Acid |
Safety and Handling
| TSCA | No |