missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrocurcumin, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014029
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014029 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014029 Supplier Merck Emd Millipore Supplier No. 5020210MG
Only null left

Tetrahydrocurcumin is a colorless compound and a major metabolite of curcumin, which is known to possess antioxidant activity and inhibitory effects on tertiary butyl hydroperoxide-induced lipid peroxidation. It can be obtained via hydrogenation of curcumin in the presence of platinum(IV)oxide.

Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 36062-04-1
Flash Point 196°C
Physical Form Neat
Linear Formula C21H24O6
MDL Number MFCD04152347
Synonym 1,7-Bis(4-hydroxy-3-methoxyphenyl)-3,5-heptanedione
Type Analytical Standard
Quantity 10 mg
Formula Weight 372.41
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Show More Show Less