missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-beta-carboline, 98%

Catalog No. AC427580010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC427580010 1 g Glass bottle
AC427580050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC427580010 Supplier Thermo Scientific Chemicals Supplier No. 427580010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 16502-01-5 | C11H13ClN2 | 208.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16502-01-5
Molecular Formula C11H13ClN2
Molecular Weight (g/mol) 208.69
MDL Number MFCD00004954
InChI Key PHLJRXUBLWEPCM-UHFFFAOYSA-N
Synonym tryptoline, 2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, noreleagnine, 1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 1,2,3,4-tetrahydro-beta-carboline, tetrahydro-beta-carboline, thbc, tetrahydronorharman, unii-65027tmi0h, 1h,2h,3h,4h,9h-pyrido 3,4-b indole
PubChem CID 107838
IUPAC Name 2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
SMILES [H+].[Cl-].C1CC2=C(CN1)NC1=CC=CC=C21

Specifications

Melting Point 206°C to 208°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 172.23
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Tetrahydro-β-carboline
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

RUO – Research Use Only