missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-4H-thiopyran-4-one, 99%

Catalog No. AAA1587206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1587206 5 g
AAA1587203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1587206 Supplier Thermo Scientific Chemicals Supplier No. A1587206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1072-72-6 | C5H8OS | 116.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072-72-6
Molecular Formula C5H8OS
Molecular Weight (g/mol) 116.18
MDL Number MFCD00006660
InChI Key OVRJVKCZJCNSOW-UHFFFAOYSA-N
Synonym tetrahydrothiopyran-4-one, tetrahydro-4h-thiopyran-4-one, 4-oxothiane, 4-thiacyclohexanone, penthianone, 4h-thiopyran-4-one, tetrahydro, 1-thiacyclohexan-4-one, dihydro-2h-thiopyran-4 3h-one, 4-oxotetrahydrothiopyran, tetrahydro-4-thiopyrone
PubChem CID 66173
IUPAC Name thian-4-one
SMILES O=C1CCSCC1

Specifications

Melting Point 60°C to 64°C
Odor Weak
Quantity 5 g
Formula Weight 116.18
Percent Purity 99%
Chemical Name or Material Tetrahydro-4H-thiopyran-4-one
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 214-015-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only