missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2H-thiopyran-4-ol 98.0+%, TCI America™

Catalog No. T33731G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T33731G 1 g
T33735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T33731G Supplier TCI America Supplier No. T33731G

Chemical Identifiers

CAS 29683-23-6
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.19
MDL Number MFCD02942489
InChI Key YODQQARABJQLIP-UHFFFAOYSA-N
Synonym tetrahydro-2h-thiopyran-4-ol, tetrahydrothiopyran-4-ol, thiacyclohexane-4-ol, 2h-thiopyran-4-ol, tetrahydro, 4-hydroxythiane, acmc-209ydy, tetrahydrothiapyran-4-ol, 4-hydroxytetrahydrothiopyran, 4-hydroxy-tetrahydrothiopyran, tetrahydro-4h-thiopyran-4-ol
PubChem CID 576296
IUPAC Name thian-4-ol
SMILES OC1CCSCC1

Specifications

Melting Point 50°C
Color White
Boiling Point 70°C
Quantity 1 g
Formula Weight 118.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrahydro-2H-thiopyran-4-ol