missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydro-2-pyrimidinone 97.0+%, TCI America™
$126.76 - $1002.86
Chemical Identifiers
| CAS | 1852-17-1 |
|---|---|
| Molecular Formula | C4H8N2O |
| Molecular Weight (g/mol) | 100.12 |
| MDL Number | MFCD00014593 |
| InChI Key | NQPJDJVGBDHCAD-UHFFFAOYSA-N |
| PubChem CID | 74615 |
| IUPAC Name | 1,3-diazinan-2-one |
| SMILES | O=C1NCCCN1 |
Chemical Identifiers
| 1852-17-1 | |
| 100.12 | |
| NQPJDJVGBDHCAD-UHFFFAOYSA-N | |
| 1,3-diazinan-2-one |
| C4H8N2O | |
| MFCD00014593 | |
| 74615 | |
| O=C1NCCCN1 |
Specifications
| 1852-17-1 | |
| White | |
| MFCD00014593 | |
| Propyleneurea, N,N′C-Trimethyleneurea | |
| O=C1NCCCN1 | |
| 100.12 | |
| 100.12 | |
| Crystalline Powder |
| 266°C | |
| C4H8N2O | |
| 25 g | |
| NQPJDJVGBDHCAD-UHFFFAOYSA-N | |
| 1,3-diazinan-2-one | |
| 74615 | |
| ≥97.0% (T) | |
| Tetrahydro-2-pyrimidinone |
Safety and Handling
EINECSNumber : (2)-1737&(5)-0432&(5)-0945
RTECSNumber : UW7539800
TSCA : Yes