accessibility menu, dialog, popup

UserName

ONHHN

CAS RN 1852-17-1

IUPAC Name: 1,3-diazinan-2-one
Synonyms: 1,3-diazinan-2-one
Molecular Weight (g/mol): 100.12
Molecular Formula: C4H8N2O
InChi Key: NQPJDJVGBDHCAD-UHFFFAOYSA-N
SMILES: O=C1NCCCN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results