missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2-pyrimidinone 97.0+%, TCI America™

Catalog No. P052625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P052625G 25 g
P0526500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P052625G Supplier TCI America Supplier No. P052625G
Only null left

Chemical Identifiers

CAS 1852-17-1
Molecular Formula C4H8N2O
Molecular Weight (g/mol) 100.12
MDL Number MFCD00014593
InChI Key NQPJDJVGBDHCAD-UHFFFAOYSA-N
PubChem CID 74615
IUPAC Name 1,3-diazinan-2-one
SMILES O=C1NCCCN1

Specifications

Melting Point 266°C
Color White
Quantity 25 g
Synonym Propyleneurea, N,N′C-Trimethyleneurea
Formula Weight 100.12
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrahydro-2-pyrimidinone
EINECSNumber (2)-1737&(5)-0432&(5)-0945
RTECSNumber UW7539800
TSCA Yes