missing translation for 'onlineSavingsMsg'
Learn More

Tetraheptylammonium Bromide 98.0+%, TCI America™

Supplier:  TCI America T160225G

 View more versions of this product

Catalog No. T160225G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetraheptylammonium Bromide
89°C
C28H60BrN
25 g
YQIVQBMEBZGFBY-UHFFFAOYSA-M
tetraheptylazanium;bromide
78073
≥98.0% (T)
4368-51-8
White
MFCD00011861
tetraheptylammonium bromide, tetra-n-heptylammonium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide, tetraheptylazanium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide 1:1, acmc-209jv6, nhep4 1+ *br 1-
CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Br-]
490.699
490.70
Crystalline Powder

Chemical Identifiers

4368-51-8
490.699
YQIVQBMEBZGFBY-UHFFFAOYSA-M
78073
CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Br-]
C28H60BrN
MFCD00011861
tetraheptylammonium bromide, tetra-n-heptylammonium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide, tetraheptylazanium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide 1:1, acmc-209jv6, nhep4 1+ *br 1-
tetraheptylazanium;bromide
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions