missing translation for 'onlineSavingsMsg'
Learn More

Tetraheptylammonium Bromide 98.0+%, TCI America™

Catalog No. p-7136867
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16025G 5 g
T160225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16025G Supplier TCI America Supplier No. T16025G
Only null left

Chemical Identifiers

CAS 4368-51-8
Molecular Formula C28H60BrN
Molecular Weight (g/mol) 490.699
MDL Number MFCD00011861
InChI Key YQIVQBMEBZGFBY-UHFFFAOYSA-M
Synonym tetraheptylammonium bromide, tetra-n-heptylammonium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide, tetraheptylazanium bromide, 1-heptanaminium, n,n,n-triheptyl-, bromide 1:1, acmc-209jv6, nhep4 1+ *br 1-
PubChem CID 78073
IUPAC Name tetraheptylazanium;bromide
SMILES CCCCCCC[N+](CCCCCCC)(CCCCCCC)CCCCCCC.[Br-]

Specifications

Melting Point 89°C
Color White
Quantity 5 g
Formula Weight 490.70
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraheptylammonium Bromide
EINECSNumber (2)-0186
TSCA Yes