missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluorosuccinic Acid 98.0+%, TCI America™

Catalog No. T16215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T16215G 5 g
T162125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T16215G Supplier TCI America Supplier No. T16215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 377-38-8
Molecular Formula C4H2F4O4
Molecular Weight (g/mol) 190.05
MDL Number MFCD00042430
InChI Key YUDUFRYTKFGQCL-UHFFFAOYSA-N
PubChem CID 67833
IUPAC Name 2,2,3,3-tetrafluorobutanedioic acid
SMILES C(=O)(C(C(C(=O)O)(F)F)(F)F)O

Specifications

Melting Point 116°C
Color White-Yellow
Boiling Point 120°C
Quantity 5 g
Synonym tetrafluorosuccinic acid, perfluorosuccinic acid, butanedioic acid, tetrafluoro, succinic acid, perfluoro, succinic acid, tetrafluoro, tetrafluorobutanedioic acid, butanedioic acid, 2,2,3,3-tetrafluoro, 2,2,3,3-tetrafluorosuccinic acid, butanedioic acid,2,2,3,3-tetrafluoro, perfluorosuccinate
Formula Weight 190.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluorosuccinic Acid
RTECSNumber WN0725000
TSCA Yes