missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluorosuccinic acid, 97%

Catalog No. AAB2046203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2046203 1 g
AAB2046206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2046203 Supplier Thermo Scientific Chemicals Supplier No. B2046203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 377-38-8 | C4H2F4O4 | 190.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 377-38-8
Molecular Formula C4H2F4O4
Molecular Weight (g/mol) 190.05
MDL Number MFCD00042430
InChI Key YUDUFRYTKFGQCL-UHFFFAOYSA-N
PubChem CID 67833
IUPAC Name 2,2,3,3-tetrafluorobutanedioic acid
SMILES C(=O)(C(C(C(=O)O)(F)F)(F)F)O

Specifications

Melting Point ∼115°C
Density 1.4
Boiling Point 150°C (15 mmHg)
Linear Formula HO2CCF2CF2CO2H
Quantity 1 g
UN Number UN3261
Beilstein 1790197
Synonym tetrafluorosuccinic acid, perfluorosuccinic acid, butanedioic acid, tetrafluoro, succinic acid, perfluoro, succinic acid, tetrafluoro, tetrafluorobutanedioic acid, butanedioic acid, 2,2,3,3-tetrafluoro, 2,2,3,3-tetrafluorosuccinic acid, butanedioic acid,2,2,3,3-tetrafluoro, perfluorosuccinate
Formula Weight 190.05
Percent Purity 97%
Chemical Name or Material Tetrafluorosuccinic acid
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-820-7
RTECSNumber WN0725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only