missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluoroisophthalic Acid 98.0+%, TCI America™

Catalog No. T13745G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13745G 5 g
T13741G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13745G Supplier TCI America Supplier No. T13745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1551-39-9
Molecular Formula C8F4O4
Molecular Weight (g/mol) 236.08
MDL Number MFCD00042379
InChI Key PGRIMKUYGUHAKH-UHFFFAOYSA-L
Synonym tetrafluoroisophthalic acid, 2,4,5,6-tetrafluoroisophthalic acid, tetrafluoroisophthalicacid, tetrafluorobenzene-1,3-dicarboxylic acid, tetrafluoroisophthalic, rarechem al bo 0259, acmc-1c1aw
PubChem CID 297552
IUPAC Name tetrafluorobenzene-1,3-dicarboxylate
SMILES [O-]C(=O)C1=C(F)C(C([O-])=O)=C(F)C(F)=C1F

Specifications

Melting Point 211°C
Color White
Quantity 5 g
Formula Weight 238.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluoroisophthalic Acid