accessibility menu, dialog, popup

UserName

CAS RN 1551-39-9

IUPAC Name: tetrafluorobenzene-1,3-dicarboxylate
Synonyms: tetrafluorobenzene-1,3-dicarboxylate
Molecular Weight (g/mol): 236.08
Molecular Formula: C8F4O4
InChi Key: PGRIMKUYGUHAKH-UHFFFAOYSA-L
SMILES: [O-]C(=O)C1=C(F)C(C([O-])=O)=C(F)C(F)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results