missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluorohydroquinone 97.0+%, TCI America™

Catalog No. t20035g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T20035G Supplier TCI America Supplier No. T20035G
Only null left

Chemical Identifiers

CAS 771-63-1
Molecular Formula C6H2F4O2
Molecular Weight (g/mol) 182.07
MDL Number MFCD00002340
InChI Key ZSDAMBJDFDRLSS-UHFFFAOYSA-N
Synonym tetrafluorobenzene-1,4-diol, tetrafluorohydroquinone, 2,3,5,6-tetrafluorohydroquinone, hydroquinone, tetrafluoro, tetrafluoro-1,4-hydroquinone, 1,4-benzenediol, 2,3,5,6-tetrafluoro, 1,2,4,5-tetrafluoro-3,6-dihydroxybenzene, 1,4-dihydroxy-2,3,5,6-tetrafluorobenzene, tetrafluoro-1,4-benzenediol
PubChem CID 69870
IUPAC Name tetrafluorobenzene-1,4-diol
SMILES OC1=C(F)C(F)=C(O)C(F)=C1F

Specifications

Melting Point 175°C
Quantity 5 g
Formula Weight 182.07
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tetrafluorohydroquinone