accessibility menu, dialog, popup

UserName

CAS RN 771-63-1

IUPAC Name: tetrafluorobenzene-1,4-diol
Synonyms: tetrafluorobenzene-1,4-diol
Molecular Weight (g/mol): 182.07
Molecular Formula: C6H2F4O2
InChi Key: ZSDAMBJDFDRLSS-UHFFFAOYSA-N
SMILES: OC1=C(F)C(F)=C(O)C(F)=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results