missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylthiuram Disulfide 97.0+%, TCI America™
Supplier: TCI America B0479500G
Specifications
| Tetraethylthiuram Disulfide | |
| 70°C | |
| C10H20N2S4 | |
| 500 g | |
| disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd | |
| CCN(CC)C(=S)SSC(=S)N(CC)CC | |
| 296.524 | |
| CHEBI:4659 | |
| ≥97.0% (T) |
| 97-77-8 | |
| White-Yellow | |
| MFCD00009048 | |
| 3077 | |
| AUZONCFQVSMFAP-UHFFFAOYSA-N | |
| diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate | |
| 3117 | |
| 296.52 | |
| Crystalline Powder |
Chemical Identifiers
| 97-77-8 | |
| 296.524 | |
| AUZONCFQVSMFAP-UHFFFAOYSA-N | |
| 3117 | |
| diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate |
| C10H20N2S4 | |
| MFCD00009048 | |
| disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd | |
| CHEBI:4659 | |
| CCN(CC)C(=S)SSC(=S)N(CC)CC |
Safety and Handling
EINECSNumber : (2)-1820
RTECSNumber : JO1225000
TSCA : Yes