missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylthiuram Disulfide 97.0+%, TCI America™

Catalog No. B047925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B047925G 25 g
B0479500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B047925G Supplier TCI America Supplier No. B047925G

Chemical Identifiers

CAS 97-77-8
Molecular Formula C10H20N2S4
Molecular Weight (g/mol) 296.524
MDL Number MFCD00009048
InChI Key AUZONCFQVSMFAP-UHFFFAOYSA-N
Synonym disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd
PubChem CID 3117
ChEBI CHEBI:4659
IUPAC Name diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC

Specifications

Melting Point 70°C
Color White-Yellow
Quantity 25 g
UN Number 3077
Formula Weight 296.52
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylthiuram Disulfide
EINECSNumber (2)-1820
RTECSNumber JO1225000
TSCA Yes