missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ Tetraethylthiuram Disulfide Acetonitrile (TETD) Sulfurizing Reagent, 450 mL

Catalog No. 401267
Encompass
Change view
Click to view available options
Quantity:
450 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401267 450 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 401267 Supplier Applied Biosystems™ Supplier No. 401267

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Tetraethylthiuram Disulfide Acetonitrile (TETD) Sulfurizing Reagent is used for making phosphorothiate backbone oligonucleotides.

Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
  • Reagent concentrations are within our strict specifications, guaranteeing consistent quality.
  • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product.
  • Reagent kits are available for your convenience, and save you time and money.

Specifications

Packaging Type Shipped at Room Temperature
Quantity 450 mL

For Research Use Only. Not for use in diagnostic procedures.