missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylthiuram Disulfide, 97%

CAS: 97-77-8 | C10H20N2S4 | 296.51 g/mol

$62.56 - $62.56

Chemical Identifiers

CAS 97-77-8
Molecular Formula C10H20N2S4
Molecular Weight (g/mol) 296.51
MDL Number MFCD00008157
InChI Key AUZONCFQVSMFAP-UHFFFAOYSA-N
Synonym disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd
PubChem CID 3117
ChEBI CHEBI:4659
IUPAC Name diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC138120250
SDS View Documents
Thermo Scientific Chemicals
138120250
25 g Plastic bottle
Each for $62.56
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

97-77-8
296.51
AUZONCFQVSMFAP-UHFFFAOYSA-N
3117
diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
C10H20N2S4
MFCD00008157
disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd
CHEBI:4659
CCN(CC)C(=S)SSC(=S)N(CC)CC

Specifications

97-77-8
100.0
Yellow
0.3% max. (60°C)
96% min. (HPLC)
C10H20N2S4
MFCD00008157
04, 122
15, 3407
Solubility in water: 0.02g/100mL. Other solubilities: 3.82g/100mL alcohol, 7.14g/100mL ether, soluble in acetone, benzene, carbon disulfice and, chloroform
CCN(CC)C(=S)SSC(=S)N(CC)CC
296.51
CHEBI:4659
97%
Tetraethylthiuram disulfide, 95%
96.0
70.0°C to 74.0°C
117°C (23.0 hPa)
Authentic
Plastic bottle
[(C2H5)2NCS2-]2
25 g
06,569
disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd
AUZONCFQVSMFAP-UHFFFAOYSA-N
diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
3117
296.51
Crystalline Powder, Crystals or Granules
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-607-8

RTECSNumber : JO1225000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only