Learn More
Tetraethylthiuram disulfide, 97%
CAS: 97-77-8 | C10H20N2S4 | 296.524 g/mol
$92.32 - $910.90
Chemical Identifiers
| CAS | 97-77-8 |
|---|---|
| Molecular Formula | C10H20N2S4 |
| Molecular Weight (g/mol) | 296.524 |
| MDL Number | MFCD00009048 |
| InChI Key | AUZONCFQVSMFAP-UHFFFAOYSA-N |
| Synonym | disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd |
| PubChem CID | 3117 |
| ChEBI | CHEBI:4659 |
| IUPAC Name | diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate |
| SMILES | CCN(CC)C(=S)SSC(=S)N(CC)CC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2072130
|
Thermo Scientific Chemicals
B2072130 |
250 g |
Each for $92.32
|
|
|||||
|
AAB207210B
|
Thermo Scientific Chemicals
B207210B |
1000 g |
Each for $247.14
|
|
|||||
|
AAB207210I
|
Thermo Scientific Chemicals
B207210I |
5000 g |
Each for $910.90
|
|
|||||
Description
Dopamine beta-hydroxylase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 97-77-8 | |
| 296.524 | |
| AUZONCFQVSMFAP-UHFFFAOYSA-N | |
| 3117 | |
| diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate |
| C10H20N2S4 | |
| MFCD00009048 | |
| disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd | |
| CHEBI:4659 | |
| CCN(CC)C(=S)SSC(=S)N(CC)CC |
Specifications
| 97-77-8 | |
| 1.3 | |
| C10H20N2S4 | |
| 250 g | |
| 1712560 | |
| disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd | |
| CCN(CC)C(=S)SSC(=S)N(CC)CC | |
| 296.524 | |
| CHEBI:4659 | |
| 97% |
| 69°C to 72°C | |
| 117°C (17 mmHg) | |
| MFCD00009048 | |
| UN3077 | |
| 14,3364 | |
| AUZONCFQVSMFAP-UHFFFAOYSA-N | |
| diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate | |
| 3117 | |
| 296.54 | |
| Tetraethylthiuram disulfide |
Safety and Handling
GHS H Statement
H373-H302-H317
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.
P260-P264b-P270-P272-P280g-P301+P312-P302+P352-P314-P330-P333+P313-P363-P501c
H302-H317-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 202-607-8
RTECSNumber : JO1225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only