missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylsilane 97.0+%, TCI America™
Supplier: TCI America T155225ML
Specifications
| Tetraethylsilane | |
| Colorless | |
| C8H20Si | |
| 25 mL | |
| silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si | |
| CC[Si](CC)(CC)CC | |
| 144.333 | |
| 144.33 | |
| Liquid |
| 631-36-7 | |
| 155°C | |
| MFCD00009019 | |
| 1993 | |
| VCZQFJFZMMALHB-UHFFFAOYSA-N | |
| tetraethylsilane | |
| 12426 | |
| ≥97.0% (GC) |
Chemical Identifiers
| 631-36-7 | |
| 144.333 | |
| VCZQFJFZMMALHB-UHFFFAOYSA-N | |
| 12426 | |
| CC[Si](CC)(CC)CC |
| C8H20Si | |
| MFCD00009019 | |
| silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si | |
| tetraethylsilane |
Safety and Handling
RTECSNumber : VV5500000
TSCA : Yes