missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylsilane, 97%

Catalog No. AAL0332814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0332814 25 g
AAL0332822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0332814 Supplier Thermo Scientific Chemicals Supplier No. L0332814
Only null left

CAS: 631-36-7 | C8H20Si | 144.333 g/mol

Used as the intermediates of OLED, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as the intermediates of OLED, pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 631-36-7
Molecular Formula C8H20Si
Molecular Weight (g/mol) 144.333
MDL Number MFCD00009019
InChI Key VCZQFJFZMMALHB-UHFFFAOYSA-N
Synonym silane, tetraethyl, tetraethylsilicon, tetraethylsilane, acmc-209ndb, c2h5 4si
PubChem CID 12426
IUPAC Name tetraethylsilane
SMILES CC[Si](CC)(CC)CC

Specifications

Boiling Point 153°C to 155°C
Melting Point -82°C
Quantity 25 g
UN Number UN1993
Beilstein 1732768
Flash Point 26°C (78°F)
Solubility Information Insoluble in water.
Formula Weight 144.33
Percent Purity 97%
Odor Recognizable
Refractive Index 1.426
Density 0.767
Chemical Name or Material Tetraethylsilane
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 211-155-0
RTECSNumber VV5500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only