missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylenepentamine, technical

Catalog No. p-3830336
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 kg
250 g
500 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC138112500 250 g Glass bottle
AC138115000 500 g Glass bottle
AC138110025 2.5 kg Glass bottle
3 options

Catalog No. AC138112500

Supplier: Thermo Scientific Chemicals 138112500

Only null left

CAS: 112-57-2 | C8H23N5 | 189.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-57-2
Molecular Formula C8H23N5
Molecular Weight (g/mol) 189.31
MDL Number MFCD00008168
InChI Key FAGUFWYHJQFNRV-UHFFFAOYSA-N
Synonym tetraethylenepentamine, tetren, 1,4,7,10,13-pentaazatridecane, tetraethylene pentamine, tetraethylpentylamine, 1,11-diamino-3,6,9-triazaundecane, 3,6,9-triazaundecamethylenediamine, 3,6,9-triazaundecane-1,11-diamine, deh 26, unii-yzd1c9kq28
PubChem CID 8197
ChEBI CHEBI:49798
IUPAC Name N'-[2-[2-(2-aminoethylamino)ethylamino]ethyl]ethane-1,2-diamine
SMILES NCCNCCNCCNCCN

Specifications

Melting Point -40°C
Density 0.9900g/mL
Boiling Point 340°C
Flash Point 139°C
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5055
Linear Formula HN(CH2CH2NHCH2CH2NH2)2
Quantity 250 g
Specific Gravity 0.99
Solubility Information Solubility in water: soluble
Formula Weight 189.3
Percent Purity ≥95.0%
Grade Technical
Physical Form Liquid
Chemical Name or Material Tetraethylenepentamine, tech.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit
EINECSNumber 203-986-2
RTECSNumber KH8585000
TSCA TSCA

RUO – Research Use Only