accessibility menu, dialog, popup

UserName

H2NNHHNNHNH2

CAS RN 112-57-2

IUPAC Name: (2-aminoethyl)[2-({2-[(2-aminoethyl)amino]ethyl}amino)ethyl]amine
Synonyms: tetraethylenepentamine tetraethylpentylamine tetren tetraethylene pentamine tetraethylenpentamin
Molecular Weight (g/mol): 189.31
Molecular Formula: C8H23N5
InChi Key: FAGUFWYHJQFNRV-UHFFFAOYSA-N
SMILES: NCCNCCNCCNCCN
Boiling Point 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)