missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylene Glycol Monomethyl Ether 98.0+%, TCI America™
Supplier: TCI America T137225G
Specifications
| Tetraethylene Glycol Monomethyl Ether | |
| Yellow | |
| C9H20O5 | |
| 25 g | |
| ZNYRFEPBTVGZDN-UHFFFAOYSA-N | |
| 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethanol | |
| 90263 | |
| ≥98.0% (GC) |
| 23783-42-8 | |
| 125°C | |
| MFCD00041756 | |
| 2,5,8,11-tetraoxatridecan-13-ol, tetraethylene glycol monomethyl ether, tetraethyleneglycol monomethyl ether, 3,6,9,12-tetraoxatridecan-1-ol, methyl tetraglycol, tetraethyleneglycol monomethylether, mpeg4-oh, 2-2-2-2-methoxy-ethoxy-ethoxy-ethoxy-ethanol, 2-2-2-2-methoxyethoxy ethoxy ethoxy ethanol, m-peg4-alcohol | |
| COCCOCCOCCOCCO | |
| 208.254 | |
| 208.25 | |
| Liquid |
Chemical Identifiers
| 23783-42-8 | |
| 208.254 | |
| ZNYRFEPBTVGZDN-UHFFFAOYSA-N | |
| 90263 | |
| COCCOCCOCCOCCO |
| C9H20O5 | |
| MFCD00041756 | |
| 2,5,8,11-tetraoxatridecan-13-ol, tetraethylene glycol monomethyl ether, tetraethyleneglycol monomethyl ether, 3,6,9,12-tetraoxatridecan-1-ol, methyl tetraglycol, tetraethyleneglycol monomethylether, mpeg4-oh, 2-2-2-2-methoxy-ethoxy-ethoxy-ethoxy-ethanol, 2-2-2-2-methoxyethoxy ethoxy ethoxy ethanol, m-peg4-alcohol | |
| 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethanol |
Safety and Handling
EINECSNumber : (7)-0097
TSCA : Yes