missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene Glycol Monomethyl Ether 98.0+%, TCI America™

Catalog No. p-7136741
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13725G 5 g
T137225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13725G Supplier TCI America Supplier No. T13725G
Only null left

Chemical Identifiers

CAS 23783-42-8
Molecular Formula C9H20O5
Molecular Weight (g/mol) 208.254
MDL Number MFCD00041756
InChI Key ZNYRFEPBTVGZDN-UHFFFAOYSA-N
Synonym 2,5,8,11-tetraoxatridecan-13-ol, tetraethylene glycol monomethyl ether, tetraethyleneglycol monomethyl ether, 3,6,9,12-tetraoxatridecan-1-ol, methyl tetraglycol, tetraethyleneglycol monomethylether, mpeg4-oh, 2-2-2-2-methoxy-ethoxy-ethoxy-ethoxy-ethanol, 2-2-2-2-methoxyethoxy ethoxy ethoxy ethanol, m-peg4-alcohol
PubChem CID 90263
IUPAC Name 2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethanol
SMILES COCCOCCOCCOCCO

Specifications

Color Yellow
Boiling Point 125°C
Quantity 5 g
Formula Weight 208.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraethylene Glycol Monomethyl Ether
EINECSNumber (7)-0097
TSCA Yes