missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylene Glycol Dimethyl Ether 98.0+%, TCI America™

Catalog No. B049725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B049725ML 25 mL
B0497500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B049725ML Supplier TCI America Supplier No. B049725ML

Chemical Identifiers

CAS 143-24-8
Molecular Formula C10H22O5
Molecular Weight (g/mol) 222.28
MDL Number MFCD00008505
InChI Key ZUHZGEOKBKGPSW-UHFFFAOYSA-N
Synonym tetraglyme, tetraethylene glycol dimethyl ether, 2,5,8,11,14-pentaoxapentadecane, dimethoxytetraglycol, glyme 5, ansul ether 181at, dimethoxytetraethylene glycol, nissan uniox mm 200, bis 2-2-methoxyethoxy ethyl ether, methyltetraglyme200
PubChem CID 8925
ChEBI CHEBI:46785
IUPAC Name 2,5,8,11,14-pentaoxapentadecane
SMILES COCCOCCOCCOCCOC

Specifications

Color Colorless
Boiling Point 276°C
Quantity 25 mL
Formula Weight 222.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tetraethylene Glycol Dimethyl Ether
EINECSNumber (7)-1321
RTECSNumber SB0400000
TSCA Yes