accessibility menu, dialog, popup

UserName

H3COOOOOCH3

CAS RN 143-24-8

IUPAC Name: 2,5,8,11,14-pentaoxapentadecane
Synonyms: tetraglyme dimethoxytetraglycol nissan uniox MM 200 dimethoxytetraethylene glycol glyme 5 methyltetraglyme200 ansul ether 181AT
Molecular Weight (g/mol): 222.28
Molecular Formula: C10H22O5
InChi Key: ZUHZGEOKBKGPSW-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOC
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)
  • (7)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)