missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Iodide 98.0+%, TCI America™

Catalog No. p-7136078
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T009725G 25 g
T0097100G 100 g
T0097500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T009725G Supplier TCI America Supplier No. T009725G

Chemical Identifiers

CAS 68-05-3
Molecular Formula C8H20IN
Molecular Weight (g/mol) 257.16
MDL Number MFCD00011829
InChI Key UQFSVBXCNGCBBW-UHFFFAOYSA-M
Synonym tetraethylammonium iodide, tetamon iodide, tetramon j, ethanaminium, n,n,n-triethyl-, iodide, n,n,n-triethylethanaminium iodide, ammonium, tetraethyl-, iodide, teai, tetraethylammoniumiodide, tetraethylazanium iodide, tetraethyl ammonium iodide
PubChem CID 6225
IUPAC Name tetraethylazanium iodide
SMILES [I-].CC[N+](CC)(CC)CC

Specifications

Color White
Quantity 25 g
Formula Weight 257.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylammonium Iodide
EINECSNumber (2)-0186
RTECSNumber BS7365000
TSCA Yes