accessibility menu, dialog, popup

UserName

ICH3NH3CH3CH3C

CAS RN 68-05-3

IUPAC Name: tetraethylazanium iodide
Synonyms: tetraethylammonium iodide tetraethyl ammonium iodide
Molecular Weight (g/mol): 257.16
Molecular Formula: C8H20IN
InChi Key: UQFSVBXCNGCBBW-UHFFFAOYSA-M
SMILES: [I-].CC[N+](CC)(CC)CC
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)