missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Hydroxide, 10% in Water

Catalog No. AC420281000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC420281000 100 g
AC420285000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC420281000 Supplier Thermo Scientific Chemicals Supplier No. 420281000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 77-98-5 | C8H21NO | 147.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst

Specifications

Chemical Name or Material Tetraethylammonium hydroxide
Color Colorless
Name Note 10% in Water
CAS 77-98-5, 7732-18-5
Boiling Point 102.0°C
Molecular Formula C8H21NO
Linear Formula (C2H5)4NOH
MDL Number MFCD00009024
Density 1.0200g/mL
Merck Index 15, 9346
Synonym tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
Solubility Information Solubility in water: soluble.
InChI Key LRGJRHZIDJQFCL-UHFFFAOYSA-M
SMILES [OH-].CC[N+](CC)(CC)CC
IUPAC Name tetraethylazanium;hydroxide
Molecular Weight (g/mol) 147.26
PubChem CID 6509
Formula Weight 147.26
Percent Purity 9.5 to 10.5%
Packaging Plastic bottle
Refractive Index 1.402
Specific Gravity 1.02
Quantity 100 g
Physical Form Liquid
Show More Show Less

RUO – Research Use Only