missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Fluoride Trihydrofluoride 97.0+%, TCI America™

Catalog No. T202610G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T202610G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T202610G Supplier TCI America Supplier No. T202610G

Chemical Identifiers

CAS 42539-97-9
Molecular Formula C8H23F4N
Molecular Weight (g/mol) 209.273
MDL Number MFCD06797183
InChI Key HVDAPPODKZABKP-UHFFFAOYSA-M
PubChem CID 11085061
IUPAC Name tetraethylazanium;fluoride;trihydrofluoride
SMILES CC[N+](CC)(CC)CC.F.F.F.[F-]

Specifications

Color White-Yellow
Quantity 10 g
UN Number 2923
Formula Weight 209.27
Percent Purity ≥97.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Tetraethylammonium Fluoride Trihydrofluoride