accessibility menu, dialog, popup

UserName

CH3NH3CH3CH3CHFHFHFF

CAS RN 42539-97-9

IUPAC Name: tetraethylazanium trihydrofluoride fluoride
Synonyms: tetraethylammonium trihydrofluoride fluoride
Molecular Weight (g/mol): 209.27
Molecular Formula: C8H23F4N
InChi Key: HVDAPPODKZABKP-UHFFFAOYSA-M
SMILES: F.F.F.[F-].CC[N+](CC)(CC)CC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results