missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Fluoride Hydrate 98.0+%, TCI America™

Supplier:  TCI America T08385G

 View more versions of this product

Catalog No. T08385G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetraethylammonium Fluoride Hydrate
White
MFCD00011826
QSUJAUYJBJRLKV-UHFFFAOYSA-M
tetraethylazanium;fluoride
69582
≥98.0% (T)
665-46-3
C8H20FN
5 g
CC[N+](CC)(CC)CC.[F-]
149.253
149.25
Crystalline Powder

Chemical Identifiers

665-46-3
149.253
QSUJAUYJBJRLKV-UHFFFAOYSA-M
tetraethylazanium;fluoride
C8H20FN
MFCD00011826
69582
CC[N+](CC)(CC)CC.[F-]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions